Selenium chloride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Selenium(I) chloride
ampouled under argon
123320-25G
BT123320-25GEA
322
USD
BT123320-25G
Selenium chloride ≥99%
Selenium chloride
CAS: 10025-68-0; EC No: 233-037-8; MDL No: MFCD00011226
UN No: UN2922; Haz Class: 8 (6.1); Packing Group: II
Liquid; Linear Formula: Se2Cl2; MW: 228.83
Melting Point: -85°; Boiling Point: 127° (decomposes)
Density (g/mL): 2.77
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
SeCl Boiling Pt: 127 °C (dec.) Melting Pt: –85 °C Density: 2.77 g/mL |
MDL Number:
MFCD00011226 CAS Number: 10025-68-0 EINECS: 233-037-8 UN: 2922 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...