Ammonium iodide ≥99.995%
Supplier: BeanTown Chemical
123310-100G
123310-25G
123310-5G
BT123310-100GEA
809
USD
BT123310-100G
BT123310-25G
BT123310-5G
Ammonium iodide ≥99.995%
Ammonium iodide
CAS: 12027-06-4; EC No: 234-717-7; MDL No: MFCD00011424
-4 Mesh Powder; Linear Formula: NH4I ; MW: 144.94
Melting Point: 551° (sublimes)
Hygroscopic , Light Sensitive
- Hygroscopic, Light Sensitive
- -4 Mesh Powder
Formula:
NH₄I Melting Pt: 551 °C (subl.) |
MDL Number:
MFCD00011424 CAS Number: 12027-06-4 EINECS: 234-717-7 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...