Cyclohexene ≥99%

Supplier: BeanTown Chemical

123275-100ML 123275-2.5L
BT123275-100MLEA 29.57 USD
BT123275-100ML BT123275-2.5L
Cyclohexene ≥99%
Cyclohexene

CAS: 110-83-8; EC No: 203-807-8; MDL No: MFCD00001539; RTECS: GW2500000 UN No: UN2256; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C6H10; MW: 82.15 Melting Point: -104°; Boiling Point: 83°; Flash point: -12°C (10°F) Density (g/mL): 0.811; Refractive Index: 1.446


  • RTECS: GW2500000
  • Refractive index: 1.446
  • Liquid

Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 83
Melting Pt: -104
Density: 0.811 g/mL
Flash Pt: –12 °C (10 °F)
MDL Number: MFCD00001539
CAS Number: 110-83-8
EINECS: 203-807-8
UN: 2256
ADR: 3 II
Merck Index: 13,02756

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR