Lead(II) acetate trihydrate ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate
122990-250G
122990-50G
BT122990-250GEA
257
USD
BT122990-250G
BT122990-50G
Lead(II) acetate trihydrate ≥99.995% (trace metals basis)
Lead (II) acetate trihydrate
CAS: 6080-56-4; EC No: 206-104-4; MDL No: MFCD00150023; RTECS: OF8050000
UN No: UN1616; Haz Class: 6.1; Packing Group: III
Crystalline; Linear Formula: Pb(OOCCH3)2·3H2O; MW: 379.33
Melting Point: 75° (decomposes)
- RTECS: OF8050000
- Crystalline
Formula:
(H₃CCOO)₂Pb·3H₂O Melting Pt: 75 °C (dec.) |
MDL Number:
MFCD00150023 CAS Number: 6080-56-4 EINECS: 206-104-4 UN: 1616 ADR: 6.1,III |
|
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...