Lead(II) acetate trihydrate ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

122990-250G 122990-50G
BT122990-250GEA 257 USD
BT122990-250G BT122990-50G
Lead(II) acetate trihydrate ≥99.995% (trace metals basis)
Lead (II) acetate trihydrate

CAS: 6080-56-4; EC No: 206-104-4; MDL No: MFCD00150023; RTECS: OF8050000 UN No: UN1616; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: Pb(OOCCH3)2·3H2O; MW: 379.33 Melting Point: 75° (decomposes)


  • RTECS: OF8050000
  • Crystalline

Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Boiling Pt: 280
Melting Pt: 75 (dec.)
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1 III
Merck Index: 13,05415
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR