Calcium chloride, anhydrous ≥97%
Supplier: BeanTown Chemical
122950-1KG
122950-250G
122950-5KG
BT122950-1KGEA
75.03
USD
BT122950-1KG
BT122950-250G
BT122950-5KG
Calcium chloride, anhydrous ≥97%
Calcium chloride
CAS: 10043-52-4; EC No: 233-140-8; MDL No: MFCD00010903; RTECS: EV9800000
Beads/Powder; Molecular Formula: CaCl2; MW: 110.98
Melting Point: 772°
Hygroscopic
- RTECS: EV9800000
- Hygroscopic
- Beads/Powder
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...