Formaldehyde sodium bisulfite ≥95%
Supplier: BeanTown Chemical
Synonyms:
Sodium Hydroxymethanesulphonate, Sodium Hydroxymethanesulfonate, Hydroxymethanesulfonic acid sodium salt, Hydroxymethanesulphonic acid sodium salt
122855-100G
122855-2.5KG
122855-500G
BT122855-100GEA
34
USD
BT122855-100G
BT122855-2.5KG
BT122855-500G
Formaldehyde sodium bisulfite ≥95%
Formaldehyde sodium bisulfite
CAS: 870-72-4; EC No: 212-800-9; MDL No: MFCD00044664; RTECS: PB2200000
Solid; Linear Formula: HOCH2SO3Na; MW: 134.09
Melting Point: 200° (decomposes)
Hygroscopic
- RTECS: PB2200000
- Hygroscopic
- Solid
Formula:
CH₄O₄S Melting Pt: 200 °C (dec.) |
MDL Number:
MFCD00044664 CAS Number: 870-72-4 EINECS: 212-800-9 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...