2-Butoxyethanol ≥99%
Supplier: BeanTown Chemical
122745-10L
122745-2.5L
122745-500ML
BT122745-10LEA
193
USD
BT122745-10L
BT122745-2.5L
BT122745-500ML
2-Butoxyethanol ≥99%
2-Butoxyethanol
CAS: 111-76-2; EC No: 203-905-0; MDL No: MFCD00002884; RTECS: KJ8575000
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H14O2; MW: 118.17
Melting Point: -75°; Boiling Point: 169-173°; Flash point: 67°C (153°F)
Density (g/mL): 0.902; Refractive Index: 1.419
- RTECS: KJ8575000
- Refractive index: 1.419
- Liquid
Formula:
CH₃(CH₂)₃OCH₂CH₂OH Boiling Pt: 169…173 °C Melting Pt: –75 °C Density: 0.902 g/mL Flash Pt: 67 °C (153 °F) |
MDL Number:
MFCD00002884 CAS Number: 111-76-2 EINECS: 203-905-0 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...