1-Chloro-2-methylpropane ≥98%
Supplier: BeanTown Chemical
122670-250G
122670-50G
BT122670-250GEA
150.4
USD
BT122670-250G
BT122670-50G
1-Chloro-2-methylpropane ≥98%
1-Chloro-2-methylpropane
CAS: 513-36-0; EC No: 208-157-9; MDL No: MFCD00000954
UN No: UN1127; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)2CHCH2Cl; Molecular Formula: C4H9Cl; MW: 92.57
Melting Point: -131°; Boiling Point: 68-69°; Flash point: -19°C (-2°F)
Density (g/mL): 0.883; Refractive Index: 1.398
- Refractive index: 1.398
- Liquid
Formula:
C₄H₉Cl Boiling Pt: 68…69 °C Melting Pt: –131 °C Density: 0.883 g/mL Flash Pt: –19 °C (–2 °F) |
MDL Number:
MFCD00000954 CAS Number: 513-36-0 EINECS: 208-157-9 UN: 1127 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...