Benzoyl chloride ≥99%

Supplier: BeanTown Chemical

Synonyms: Benzoic acid chloride

122665-1KG 122665-250G
BT122665-1KGEA 44.58 USD
BT122665-1KG BT122665-250G BT122665-5KG
Benzoyl chloride ≥99%
Benzoyl chloride

CAS: 98-88-4; EC No: 202-710-8; MDL No: MFCD00000653 UN No: UN1736; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C7H5ClO; MW: 140.57 Melting Point: -1°; Boiling Point: 198°; Flash point: 72°C (162°F) Density (g/mL): 1.211; Refractive Index: 1.553 Moisture Sensitive


  • Refractive index: 1.553
  • Moisture Sensitive
  • Liquid

Formula: C₇H₅ClO
Boiling Pt: 198 °C
Melting Pt: –1 °C
Density: 1.211 g/mL
Flash Pt: 72 °C (162 °F)
MDL Number: MFCD00000653
CAS Number: 98-88-4
EINECS: 202-710-8
UN: 1736
ADR: 8,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR