N4-Acetylcytosine ≥98%
Supplier: BeanTown Chemical
Synonyms:
N-(2-Oxo-1,2-dihydropyrimidin-4-yl)acetamide
122580-100G
122580-25G
122580-5G
BT122580-100GEA
374
USD
BT122580-100G
BT122580-25G
BT122580-5G
N4-Acetylcytosine ≥98%
N4-Acetylcytosine
CAS: 14631-20-0; MDL No: MFCD00134466
Powder; Molecular Formula: C6H7N3O2; MW: 153.14
Melting Point:
- Powder
Formula:
C₆H₇N₃O₂ Melting Pt: >300 °C |
MDL Number:
MFCD00134466 CAS Number: 14631-20-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...