(±)-Aspartic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Aminobutanedioic acid, DL-Aspartic acid

122535-100G 122535-2.5KG 122535-500G
BT122535-100GEA 20 USD
BT122535-100G BT122535-2.5KG BT122535-500G
(±)-Aspartic acid ≥98%
(±)-Aspartic acid

CAS: 617-45-8; EC No: 217-234-6; MDL No: MFCD00063083; RTECS: CI9097800 Solid; Linear Formula: HOOCCH2CH(NH2)COOH; Molecular Formula: C4H6NO4; MW: 133.10 Melting Point: 300°


  • RTECS: CI9097800
  • Solid

Formula: C₄H₇NO₄
MW: 133.10 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00063083
CAS Number: 617-45-8
EINECS: 217-234-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR