3,5-Di-tert-butyl-4-hydroxybenzaldehyde ≥98%

Supplier: BeanTown Chemical

122515-100G 122515-25G 122515-5G
BT122515-100GEA 247 USD
BT122515-100G BT122515-25G BT122515-5G
3,5-Di-tert-butyl-4-hydroxybenzaldehyde ≥98%
3,5-Di-tert-butyl-4-hydroxybenzaldehyde

CAS: 1620-98-0; EC No: 216-592-0; MDL No: MFCD00008826; RTECS: CU5610070 Crystalline; Molecular Formula: C15H22O2; MW: 234.33 Air Sensitive


  • RTECS: CU5610070
  • Air Sensitive
  • Crystalline

Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Boiling Pt: 332
Melting Pt: 190
MDL Number: MFCD00008826
CAS Number: 1620-98-0
EINECS: 216-592-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR