Zinc bromide, anhydrous ≥98%
Supplier: BeanTown Chemical
Synonyms:
Zinc (II) bromide
122450-100G
122450-500G
BT122450-100GEA
43.12
USD
BT122450-100G
BT122450-500G
Zinc bromide, anhydrous ≥98%
Zinc bromide
CAS: 7699-45-8; EC No: 231-718-4; MDL No: MFCD00011294; RTECS: ZH1150000
UN No: UN3260; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: ZnBr2; MW: 225.19
Melting Point: 394°; Boiling Point: 697°
Density (g/mL): 4.2; Refractive Index: 1.545
Moisture sensitive
- RTECS: ZH1150000
- Refractive index: 1.545
- Moisture sensitive
- Crystalline
Formula:
Br₂Zn Boiling Pt: 697 °C Melting Pt: 394 °C Density: 4.2 g/mL |
MDL Number:
MFCD00011294 CAS Number: 7699-45-8 EINECS: 231-718-4 UN: 3260 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...