Zinc bromide, anhydrous ≥98%

Supplier: BeanTown Chemical

Synonyms: Zinc (II) bromide

122450-100G 122450-500G
BT122450-100GEA 43.12 USD
BT122450-100G BT122450-500G
Zinc bromide, anhydrous ≥98%
Zinc bromide

CAS: 7699-45-8; EC No: 231-718-4; MDL No: MFCD00011294; RTECS: ZH1150000 UN No: UN3260; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: ZnBr2; MW: 225.19 Melting Point: 394°; Boiling Point: 697° Density (g/mL): 4.2; Refractive Index: 1.545 Moisture sensitive


  • RTECS: ZH1150000
  • Refractive index: 1.545
  • Moisture sensitive
  • Crystalline

Formula: Br₂Zn
Boiling Pt: 697 °C
Melting Pt: 394 °C
Density: 4.2 g/mL
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR