Copper(I) chloride ≥99.999%

Supplier: BeanTown Chemical

Synonyms: Cuprous chloride, Copper chloride

122405-100G 122405-25G 122405-5G
BT122405-100GEA 491 USD
BT122405-100G BT122405-25G BT122405-5G
Copper(I) chloride ≥99.999%
Copper (I) chloride

CAS: 7758-89-6; EC No: 231-842-9; MDL No: MFCD00010971; RTECS: GL6990000 UN No: UN2802; Haz Class: 8; Packing Group: III Powder; Linear Formula: CuCl; MW: 98.99 Melting Point: 430°; Boiling Point: 1490° Density (g/mL): 4.14 Air Sensitive, Moisture Sensitive


  • RTECS: GL6990000
  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1367
Melting Pt: 430
Density: 4.14 g/mL
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8 III
Merck Index: 13,02690

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR