D-(-)-Ribose ≥98%
Supplier: BeanTown Chemical
122395-10G
122395-250G
122395-50G
BT122395-10GEA
25
USD
BT122395-10G
BT122395-250G
BT122395-50G
D-(-)-Ribose ≥98%
D(-)-Ribose
CAS: 50-69-1; EC No: 200-059-4; MDL No: MFCD00135453; RTECS: VJ2275000
Powder; Molecular Formula: C5H10O5; MW: 150.13
Melting Point: 88-92°
Optical Rotation: [α]20/D -19.7°, c = 4 in water
Hygroscopic
- RTECS: VJ2275000
- Optical rotation: [α]20/D -19.7°, c = 4 in water
- Hygroscopic
- Powder
Formula:
C₅H₁₀O₅ Melting Pt: 88…92 °C |
MDL Number:
MFCD00135453 CAS Number: 50-69-1 EINECS: 200-059-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...