Chromium(III) oxide ≥99%

Supplier: BeanTown Chemical

Synonyms: Chromic oxide

122240-100G 122240-500G
BT122240-100GEA 52 USD
BT122240-100G BT122240-500G
Chromium(III) oxide ≥99%
Chromium (III) oxide

CAS: 1308-38-9; EC No: 215-160-9; MDL No: MFCD00010949; RTECS: GB6475000 Powder; Molecular Formula: Cr2O3; MW: 151.99


  • RTECS: GB6475000
  • Powder

Formula: Cr₂O₃
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR