(S)-(-)-1,2-Epoxypropane ≥99%
Supplier: BeanTown Chemical
Synonyms:
(S)-(-)-Methyloxirane, (S)-(-)-Propylene oxide, (S)-(-)-Propene oxide
122130-100G
122130-25G
122130-5G
BT122130-100GEA
792
USD
BT122130-100G
BT122130-25G
BT122130-5G
(S)-(-)-1,2-Epoxypropane ≥99%
(S)-(-)-1,2-Epoxypropane
CAS: 16088-62-3; EC No: 240-241-0; MDL No: MFCD00064312; RTECS: UJ2651000
UN No: UN1280; Haz Class: 3; Packing Group: I
Liquid; Molecular Formula: C3H6O; MW: 58.08
Boiling Point: 33-34°; Flash point: -37°C (-34°F)
Density (g/mL): 0.829; Refractive Index: 1.366
- RTECS: UJ2651000
- Refractive index: 1.366
- Liquid
Formula:
C₃H₆O Boiling Pt: 33…34 °C Density: 0.829 g/mL Flash Pt: –37 °C (–34 °F) |
MDL Number:
MFCD00064312 CAS Number: 16088-62-3 EINECS: 240-241-0 UN: 1280 ADR: 3,I |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...