Benzo[b]thiophene ≥98%

Supplier: BeanTown Chemical

Synonyms: Thianaphthene, 1-Benzothiophene

122115-100G 122115-25G 122115-5G
BT122115-100GEA 257 USD
BT122115-100G BT122115-25G BT122115-5G
Benzo[b]thiophene ≥98%
Benzo[b]thiophene

CAS: 95-15-8; EC No: 202-395-7; MDL No: MFCD00005864 UN No: UN3077; Haz Class: 9; Packing Group: III Solid; Molecular Formula: C8H6S; MW: 134.20 Melting Point: 30-33°; Boiling Point: 221-222° Density (g/mL): 1.149


  • Solid

Formula: C₈H₆S
MW: 134.2 g/mol
Boiling Pt: 221…222 °C
Melting Pt: 30…33 °C
Density: 1.149 g/mL
MDL Number: MFCD00005864
CAS Number: 95-15-8
EINECS: 202-395-7
ADR: 9 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR