1-Amino-2,2,2-trifluoroethane ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,2,2-Trifluoroethylamine

122035-1G 122035-25G 122035-5G
BT122035-1GEA 52 USD
BT122035-1G BT122035-25G BT122035-5G
1-Amino-2,2,2-trifluoroethane ≥98%
1-Amino-2,2,2-trifluoroethane

CAS: 753-90-2; EC No: 212-041-3; MDL No: MFCD00008132; RTECS: KS0175000 UN No: UN2733; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: CF3CH2NH2; Molecular Formula: C2H4F3N; MW: 99.06 Boiling Point: 36-37°; Flash point: -16°C (3°F) Density (g/mL): 1.262; Refractive Index: 1.301


  • RTECS: KS0175000
  • Refractive index: 1.301
  • Liquid

Formula: C₂H₄F₃N
MW: 99.06
Boiling Pt: 35.85
Density: 1.262 g/mL
Flash Pt: –16 °C (3 °F)
MDL Number: MFCD00008132
CAS Number: 753-90-2
EINECS: 212-041-3
UN: 2733
ADR: 3 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR