Methylhydrazine sulfate ≥98%
Supplier: BeanTown Chemical
122030-25G
122030-5G
BT122030-25GEA
186.86
USD
BT122030-25G
BT122030-5G
Methylhydrazine sulfate ≥98%
Methylhydrazine sulfate
CAS: 302-15-8; EC No: 206-115-4; MDL No: MFCD00035423; RTECS: MV7750000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Linear Formula: CH3NHNH2·H2SO4; Molecular Formula: CH8N2O4S; MW: 144.15
Melting Point: 141-142°
- RTECS: MV7750000
- Solid
Formula:
CH₆N₂ Melting Pt: 141…142 °C |
MDL Number:
MFCD00035423 CAS Number: 302-15-8 EINECS: 206-115-4 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...