L-α-Aminoadipic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid
121985-1G
121985-5G
BT121985-1GEA
79
USD
BT121985-1G
BT121985-5G
L-α-Aminoadipic acid ≥98%
L-α-Aminoadipic acid
CAS: 1118-90-7; MDL No: MFCD00002636
Powder; Molecular Formula: C6H11NO4; MW: 161.16
Melting Point: 203-205° (decomposes)
- Powder
Formula:
C₆H₁₁NO₄ MW: 161.16 g/mol Melting Pt: 203…205 °C (dec.) |
MDL Number:
MFCD00002636 CAS Number: 1118-90-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...