Docusate sodium ≥96%

Supplier: BeanTown Chemical

Synonyms: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Dioctyl sulfosuccinate sodium salt, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulfonate, Sodium dioctyl sulfosuccinate, Dioctyl sulfosuccinate, sodium salt, bis(2-Ethylhexyl) sodium sulfosuccinate

121965-100G 121965-500G
BT121965-100GEA 41.17 USD
BT121965-100G BT121965-500G
Docusate sodium ≥96%
Docusate sodium

CAS: 577-11-7; EC No: 209-406-4; MDL No: MFCD00012455; RTECS: WN0525000 Solid; Molecular Formula: C20H37NaO7S; MW: 444.55 Melting Point: 173-179° Hygroscopic


  • RTECS: WN0525000
  • Hygroscopic
  • Solid

Formula: C₂₀H₃₇NaO₇S
MW: 444.57 g/mol
Melting Pt: 155
MDL Number: MFCD00012455
CAS Number: 577-11-7
EINECS: 209-406-4
Merck Index: 12,03460

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR