Water ACS, deionized
Supplier: BeanTown Chemical
Synonyms:
Dihydrogen oxide
121925-1L
121925-4L
121925-4X1L
BT121925-1LEA
40.97
USD
BT121925-1L
BT121925-4L
BT121925-4X1L
Water ACS, deionized
Water
CAS: 7732-18-5; EC No: 231-791-2; MDL No: MFCD00011332
Liquid; Molecular Formula: H2O ; MW: 18.02
Melting Point: 0°; Boiling Point: 100°
Density (g/mL): 1.000; Refractive Index: 1.333
- Refractive index: 1.333
- Liquid
Formula:
H₂O Boiling Pt: 100 °C Melting Pt: 0 °C Density: 1.000 g/mL |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...