Paraffin wax, Solidification point: 53-57°C
Supplier: BeanTown Chemical
Synonyms:
Wax Paraffin, Paraffin wax
121820-1KG
BT121820-1KGEA
39.38
USD
BT121820-1KG
Paraffin wax, Solidification point: 53-57°C
Paraffin
CAS: 8002-74-2; EC No: 232-315-6; MDL No: MFCD00132833; RTECS: RV0350000
Solid
Melting Point: 53-57°; Boiling Point: 341-665°; Flash point: 113°C (235°F)
- RTECS: RV0350000
- Solid
Boiling Pt:
341…665 °C Melting Pt: 53…57 °C Flash Pt: 113 °C (235 °F) |
MDL Number:
MFCD00132833 CAS Number: 8002-74-2 EINECS: 232-315-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...