Phenolphthalein ACS
Supplier: BeanTown Chemical
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
121735-100G
121735-500G
BT121735-100GEA
33.66
USD
BT121735-100G
BT121735-500G
Phenolphthalein ACS
Phenolphthalein
CAS: 77-09-8; EC No: 201-004-7; MDL No: MFCD00005913; RTECS: SM8380000
Powder; Molecular Formula: C20H14O4 ; MW: 318.33
Melting Point: 261-263°
Density (g/mL): 1.299
- RTECS: SM8380000
- Powder
Formula:
C₂₀H₁₄O₄ Melting Pt: 261…263 °C Density: 1.299 g/mL |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 |
|
Specification Test Results
Clarity of solution | Passes test |
Visual transition pH | 8.0-10 |
Color | Colorless to red |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...