2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl ≥97%
Supplier: BeanTown Chemical
121725-1G
121725-5G
BT121725-1GEA
103
USD
BT121725-1G
BT121725-5G
2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl ≥97%
2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl
CAS: 102714-93-2; MDL No: MFCD11053361
Crystalline/Solid; Molecular Formula: C30H41F; MW: 420.65
Melting Point: 129-140°
- Crystalline/Solid
Formula:
C₃₀H₄₁F Melting Pt: 129…140 °C |
MDL Number:
MFCD11053361 CAS Number: 102714-93-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...