Acetyl chloride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Acetyl chloride 1M in Dichloromethane
121720-1L
121720-250ML
121720-4L
121720-50ML
BT121720-1LEA
169
USD
BT121720-1L
BT121720-250ML
BT121720-4L
BT121720-50ML
Acetyl chloride ≥99%
Acetyl chloride
CAS: 75-36-5; EC No: 200-865-6; MDL No: MFCD00000719; RTECS: AO6390000
UN No: UN1717; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: CH3COCl ; MW: 78.50
Melting Point: -112°; Boiling Point: 52°; Flash point: 5°C (41°F)
Density (g/mL): 1.104; Refractive Index: 1.389
Moisture Sensitive
- RTECS: AO6390000
- Refractive index: 1.389
- Moisture Sensitive
- Liquid
Formula:
C₂H₃ClO Boiling Pt: 52 °C Melting Pt: –112 °C Density: 1.104 g/mL Flash Pt: 5 °C (41 °F) |
MDL Number:
MFCD00000719 CAS Number: 75-36-5 EINECS: 200-865-6 UN: 1717 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...