Mercury(II) nitrate monohydrate ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: Mercury(II) nitrate hydrate, Mercury dinitrate monohydrate

121540-250G 121540-50G
BT121540-250GEA 242.65 USD
BT121540-250G BT121540-50G
Mercury(II) nitrate monohydrate ≥98.0% ACS
Mercury (II) nitrate monohydrate

CAS: 7783-34-8; EC No: 233-886-4; MDL No: MFCD00149736 UN No: UN1625; Haz Class: 6.1; Packing Group: II Crystalline; Linear Formula: Hg(NO3)2·xH2O; MW: 324.60 (anhydrous) Melting Point: 79° Density (g/mL): 4.39 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: Hg(NO₃)₂·1H₂O
MW: 342.62 g/mol
Melting Pt: 79
Density: 4.39 g/mL
MDL Number: MFCD00149736
CAS Number: 7783-34-8
EINECS: 233-886-4
UN: 1625
ADR: 6.1 II

Order Now

Specification Test Results

Assay 98.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR