Copper(II) sulfate, anhydrous
Supplier: BeanTown Chemical
Synonyms:
Cupric sulphate, Cupric sulfate
121535-100G
121535-2KG
121535-500G
BT121535-100GEA
53.77
USD
BT121535-100G
BT121535-2KG
BT121535-500G
Copper(II) sulfate, anhydrous
Copper (II) sulphate
CAS: 7758-98-7; EC No: 231-847-6; MDL No: MFCD00010981; RTECS: GL8800000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: CuSO4; MW: 159.60
Melting Point: 560° (decomposes)
Density (g/mL): 3.603
Hygroscopic
- RTECS: GL8800000
- Hygroscopic
- Powder
Formula:
CuSO₄ Melting Pt: 560 °C (dec.) Density: 3.603 g/mL |
MDL Number:
MFCD00010981 CAS Number: 7758-98-7 EINECS: 231-847-6 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...