tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Bis-Boc-pyrazolocarboxamidine, 1-[N,N'-Bis(tert-butoxycarbonyl)carbamimidoyl]-1H-pyrazole, N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, N,N'-Di-Boc-1H-pyrazole-1-carboximidamide, 1H-Pyrazole-1-carboxamidine, N,N'-Bis-BOC protected, N,N'-Bis(tert-butoxycarbonyl)-1H-pyrazole-1-carboximidamide
121395-1G
121395-25G
121395-5G
BT121395-1GEA
29
USD
BT121395-1G
BT121395-25G
BT121395-5G
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate
CAS: 152120-54-2; MDL No: MFCD01075122
Solid; Molecular Formula: C14H22N4O4 ; MW: 310.35
Melting Point: 86-90°
- Solid
Formula:
C₁₄H₂₂N₄O₄ Melting Pt: 86…90 °C |
MDL Number:
MFCD01075122 CAS Number: 152120-54-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...