Silver protein

Supplier: BeanTown Chemical

Synonyms: Silver colloidal, Silver nucleate, Silvol, Mild protargin, Colloidal Silver

121335-100G 121335-25G 121335-5G
BT121335-100GEA 290 USD
BT121335-100G BT121335-25G BT121335-5G
Silver protein
Silver protein

RTECS: UK9510000
Form: Powder
Sensitivity: Hygroscopic, Light sensitive
Merck Number: 14,8527

Formula: Ag (with protein)
MDL Number: MFCD00132268
CAS Number: 9015-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR