Naftopidil ≥98%
Supplier: BeanTown Chemical
Synonyms:
1-[4-(2-Methoxyphenyl)piperazinyl]-3-(1-naphthyloxy)propan-2-ol
121300-5G
BT121300-5GEA
230
USD
BT121300-5G
Naftopidil ≥98%
Naftopidil
CAS: 57149-07-2; MDL No: MFCD00242741; RTECS: TL9336500
Solid; Molecular Formula: C24H28N2O3; MW: 392.49
Melting Point: 127°
- RTECS: TL9336500
- Solid
Formula:
C₂₄H₂₈N₂O₃ MW: 392.50 g/mol Melting Pt: 127 °C |
MDL Number:
MFCD00242741 CAS Number: 57149-07-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...