Sulfuric acid 95.0-98.0% ACS
Supplier: BeanTown Chemical
121240-1KG
BT121240-1KGEA
53
USD
BT121240-1KG
Sulfuric acid 95.0-98.0% ACS
Sulfuric acid
CAS: 7664-93-9; EC No: 231-639-5; MDL No: MFCD00064589; RTECS: WS5600000
UN No: UN1830; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: H2SO4; MW: 98.08
Melting Point: 3°; Boiling Point: 290°
Density (g/mL): 1.84
- RTECS: WS5600000
- Liquid
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.84 g/mL |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8 II |
Specification Test Results
Assay | 95.0-98.0% |
Color (APHA) | ≤10 |
Residue after ignition | ≤5ppm |
Nitrate (NO3) | ≤0.5ppm |
Chloride (Cl) | ≤0.2ppm |
Arsenic (As) | ≤0.01ppm |
Ammonium (NH4) | ≤2ppm |
Substances reducing KMnO4 | Passes test |
Heavy metals (as Pb) | ≤1ppm |
Iron (Fe) | ≤0.2ppm |
Mercury (Hg) | ≤5ppb |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...