3-Amino-3-phenyl-1-propanol ≥98%
Supplier: BeanTown Chemical
121205-1G
121205-5G
BT121205-1GEA
74
USD
BT121205-1G
BT121205-5G
3-Amino-3-phenyl-1-propanol ≥98%
3-Amino-3-phenyl-1-propanol
CAS: 14593-04-5; EC No: 221-674-4; MDL No: MFCD00801124
Solid; Molecular Formula: C9H13NO; MW: 151.21
Melting Point: 70-77°
- Solid
Formula:
C₉H₁₃NO Melting Pt: 70…77 °C |
MDL Number:
MFCD00801124 CAS Number: 14593-04-5 EINECS: 221-674-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...