Ethyl-5-bromonicotinate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 5-bromonicotinate
121170-1G
121170-25G
121170-5G
BT121170-1GEA
50
USD
BT121170-1G
BT121170-25G
BT121170-5G
Ethyl-5-bromonicotinate ≥98%
Ethyl 5-bromonicotinate
CAS: 20986-40-7; MDL No: MFCD00040366
Solid; Molecular Formula: C8H8BrNO2; MW: 230.07
Melting Point: 39-42°; Boiling Point: 86-92°/0.5 mmHg
- Solid
Formula:
C₈H₈BrNO₂ Boiling Pt: 86…92 °C/0.5 mmHg Melting Pt: 39…42 °C |
MDL Number:
MFCD00040366 CAS Number: 20986-40-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...