Magnesium nitrate hydrate ≥99.999%
Supplier: BeanTown Chemical
|
Warning
|
121060-100G
121060-25G
121060-5G
BT121060-100GEA
249
USD
BT121060-100G
BT121060-25G
BT121060-5G
Magnesium nitrate hydrate ≥99.999%
Magnesium nitrate hydrate
CAS: 10377-60-3; EC No: 233-826-7; MDL No: MFCD00011103; RTECS: OM3756000
UN No: UN1474; Haz Class: 5.1; Packing Group: III
Crystalline Aggregates; Linear Formula: Mg(NO3)2·xH2O; MW: 148.32 (anhydrous)
Melting Point: 89°; Boiling Point: 330° (decomposes)
Density (g/mL): 1.46
Hygroscopic
- RTECS: OM3756000
- Crystalline Aggregates
- Hygroscopic
Formula:
Mg(NO₃)₂·xH₂O MW: 148.32 g/mol (anhy) Boiling Pt: 330 °C (dec.) Melting Pt: 89 °C Density: 1.46 g/mL |
MDL Number:
MFCD00011103 CAS Number: 10377-60-3 EINECS: 233-826-7 UN: 1474 ADR: 5.1 III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...