Pentafluorophenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,3,4,5,6-Pentafluorobenzeneboronic acid, Dihydroxy(pentafluorophenyl)borane, (Pentafluorophenyl)metaboric acid, Pentafluorobenzeneboronic acid

121055-100G 121055-25G 121055-5G
BT121055-100GEA 1134 USD
BT121055-100G BT121055-25G BT121055-5G
Pentafluorophenylboronic acid ≥97%
Pentafluorophenylboronic acid

CAS: 1582-24-7; EC No: 000-000-0; MDL No: MFCD01074663 Solid; Molecular Formula: C6H2BF5O2; MW: 211.88 Moisture Sensitive


  • Moisture Sensitive
  • Solid

Formula: C₆H₂BF₅O₂
MDL Number: MFCD01074663
CAS Number: 1582-24-7

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR