nanoVette Microliter Measurement Cell, Beckman Coulter®
Supplier: Beckman Coulter
A44100
BKA44097EA 6375.13 USD
BKA44097 BKA44098 BKA44100
nanoVette Microliter Measurement Cell, Beckman Coulter®
Cuvettes Spectrophotometer Cuvettes
The nanoVette fiber-optic measurement cell fits into the cuvette holders of Beckman Coulter® spectrophotometers for sub-microliter analysis in DNA and protein applications.
  • For Measurement of Nucleic Acid and Protein Concentrations
  • No Dilution Required
  • Minimal Preparation
  • Interchangeable Lids
  • Reusable Samples
It measures nucleic acid—dsDNA, ssDNA, RNA, and Oligo—or protein concentrations in the sub-microliter range by reducing the optical path length to 1.0 or 0.2 mm via interchangeable lids.
No dilution is required, so preparation is minimal, and samples can be reused downstream. The cell can be removed and reinserted with ease depending upon sample size, allowing for maximum flexibility with spectrophotometer function.