Ammonium Ion Selective Combination Electrode, Beckman Coulter®
Supplier: Beckman Coulter
511142
BK511142EA 1024.09 USD
BK511142
Ammonium Ion Selective Combination Electrode, Beckman Coulter®
Electrodes ISE Electrodes
This combination electrode comes with a protective epoxy body and develops a potential at the membrane surface that is related to the specific ion.
  • Application: Developing Potential at the Membrane Surface
  • Refillable Epoxy Body Construction
  • Filling Solution (15 mL)
  • One-Meter Cable with BNC Connector
  • Polymer Membrane Sensor
It is supplied with an attached, one-meter cable with BNC connector and 15 mL of filling solution.
Length/useable length: 140/95 mm