JA-10 Fixed-Angle Rotor, Beckman Coulter®
Supplier: Beckman Coulter
369687
BK369687EA 12795.95 USD
BK369687
JA-10 Fixed-Angle Rotor, Beckman Coulter®
Centrifuges Rotors
Six-place, aluminum rotors are for use with Avanti™ centrifuges.
  • Ideal for Initial Processing of Tissue Homogenates and Other Large Particles
  • Carrying Handle Included
  • Accommodate Six 500 mL Bottles
Large-volume rotors are ideal for initial processing of tissue homogenates and other large particles. They accommodate six 500 mL bottles and include a carrying handle.