S5700 Swinging Bucket Rotor, Beckman Coulter®
Supplier: Beckman Coulter
368954
BK368954EA 5676.65 USD
BK368954
S5700 Swinging Bucket Rotor, Beckman Coulter®
Centrifuges Rotors
These rotors are for use with the Allegra® 25R centrifuge.
  • Applications: DNA isolation and purification
  • Material: Aluminum
  • Maximum Speed: 5700 rpm
  • Maximum Capacity: 2 x 5 Plates
  • Microplate Carriers with Support Pads Included
They provide DNA isolation/purification and post-reaction cleanup in the 96-well format
The rotors can also hold standard microplates used in the serial dilution of small liquid volumes: up to five stacked 96-well polypropylene plates, two stacked deep-well plates, or one square-well plate per bucket.
Each rotor bucket can carry a 96-well kit (DNA or RNA) for high throughput processing.
Ordering information: Rotor supplied with two removable buckets and microplate carrier kits.