TS-5.1-500 Swinging Bucket Rotor, Beckman Coulter®
Supplier: Beckman Coulter
368308
BK368308EA 5275.88 USD
BK368308
TS-5.1-500 Swinging Bucket Rotor, Beckman Coulter®
Centrifuges Rotors
This Rotor is for use with the Allegra® 25R centrifuge.
  • Four Removable Buckets
  • Versatile
  • Horizontal Rotor
  • Ideal for sedimenting protein precipitates, large cell particles, and cell debris in a wide range of volumes
This horizontal rotor is ideal for sedimenting protein precipitates, large cell particles, and cell debris in a wide range of volumes.
It accepts 0.5 to 500 mL per tube up to 2 L and 16x50 mL and 36x15 mL conical tubes for high capacity pelleting in conical labware. For DNA precipitation and other microplate applications, a microplate carrier is available.
Certifications: Optional bucket covers are certified by Porton Down, UK for biocontainment.
Accessories information: Each microplate carrier can hold up to two stacked 96-well microplates or one deep-well plate.
Ordering information: Rotor supplied with four removable buckets. Covers and adapters sold separately.