Allegra® 64R High-Speed Centrifuge, Beckman Coulter®
Supplier: Beckman Coulter
367586
BK367586EA 21245.6 USD
BK367586
Allegra® 64R High-Speed Centrifuge, Beckman Coulter®
Centrifuges Benchtop Centrifuges
This brushless induction drive refrigerated centrifuge performs routine spins on the lab bench.
  • Ideal for High-Speed Separations, Including Cell Gradient Spins and Nucleic Acid Precipitations
  • Easy-To-Use Control Pane
  • Customizable for Routine Runs with Multiple Acceleration and Deceleration Profiles
  • Door Interlock
  • Over-Temperature/Over-Speed Diagnostics
The quiet (65 dB), brushless induction drive motor can be set in 100 rpm increments and is accurate to ±50rpm
An easy-to-use control panel displays actual speed, RCF, time, temperature, and acceleration and deceleration rates. It can be customized for routine runs with multiple acceleration and deceleration profiles. Safety features include a door interlock that prevents contact with spinning rotor and over-temperature and over-speed diagnostics that protect samples. Refrigerant is free of CFC. The biosafe bowl rotor contains hazardous spills and aerosols if breakage occurs. It accommodates up to 84 microcentrifuge tubes from 250 µL to 1.8 mL and can be carried from the centrifuge to a fume hood without breaking the seal.
A flexible drive shaft helps compensate for drive imbalances. It accommodates a selection of 13 rotors, sold separately.
Certifications: CSA certified. UL listed. C-UL listed. CE marked.
Ordering information: Supplied with a manufacturer’s one-year parts and labor warranty. Rotors and accessories sold separately.