JS-4.3 Swinging Bucket Rotor, Beckman Coulter®
Supplier: Beckman Coulter
362734
BK362734EA 10307.68 USD
BK362734
JS-4.3 Swinging Bucket Rotor, Beckman Coulter®
Centrifuges Rotors
Four-place swinging bucket rotors with anodized, aluminum buckets are designed for use in Avanti™ J-20XP and JXPIBK393124 series centrifuges.
  • Ideal for Rapid Sedimentation of Protein Precipitates, Large Particles, Cells, and Cell Debris
  • Other Applications: Binding Studies and Separating Serum from Whole Blood
  • Space for Four 750 mL Tubes
  • Anodized Aluminum Buckets
  • Four-Place Rotors
These rotors are ideal for rapid sedimentation of protein precipitates, large particles, cells, and cell debris
They accommodate four 750 mL tubes.
They can also be used for binding studies and for separating serum from whole blood.
Ordering information: Accessories include modular disk adapters (BK356968 series), Aerosolve® canisters (BK359481 series), and bottle sleeves (BK349946 series). The rotor buckets are interchangeable with MicroPlus™ microplate carriers (BK362394 series).