Polypropylene Bottle with Screw-On Cap
Supplier: Beckman Coulter
356855
BK356855PK 158.69 USD
BK356855
Polypropylene Bottle with Screw-On Cap
Bottles Plastic Bottles
Bottles are supplied with screw-on caps for use in benchtop centrifuges and microcentrifuges rotors.
  • Polypropylene construction
  • Screw-on closures included
  • Nominal capacity: 750 ml
Bottles are manufactured under ISO 9001 guidelines.