Specialty Adapter for GH-3.8/SX4750 Rotor, Beckman Coulter
Supplier: Beckman Coulter
354500
BK354500PK 2325.36 USD
BK354500
Specialty Adapter for GH-3.8/SX4750 Rotor, Beckman Coulter
Centrifuges Rotors
This specialty adapter is for use with the following products: GH-3.8 (BK360581) SX4750 (BK369702), Allegra X-15R (BK392932), Allegra X-14 (BKA99465 or BKA99464), and Allegra 6 (BK366802).
  • Adapter accommodates a wide variety of tubes
  • For use with 100 ml, short-form conical oil tubes
GH-3.8A rotor accommodates tubes with a maximum length of 120 mm.
Safely spins 100 ml short-form conical oil tubes (44 x 165 mm).
100 ml adapters are not compatible with the GH-3.8A rotor.
Delivery information: Adapter is supplied with a pad.