Thickwall Polycarbonate Tubes, Beckman Coulter
Supplier: Beckman Coulter
349622
BK349622PK 292.45 USD
BK349622
Thickwall Polycarbonate Tubes, Beckman Coulter
Tubes Centrifuge Tubes
Thickwall polycarbonate tubes are for use in ultracentrifuge rotors.
They have an open-top, thickwall closure type and are transparent. Made of polycarbonate material.