Airfuge® Ultracentrifuges, Beckman Coulter®
Supplier: Beckman Coulter
340400
BK340400EA 18161.4 USD
BK340400
Airfuge® Ultracentrifuges, Beckman Coulter®
Centrifuges Microcentrifuges
This compact ultracentrifuge is a versatile and convenient benchtop unit. A variety of rotors make the Airfuge ideal for applications such as receptor-binding studies, lipoprotein assays, chylomicron studies, sample preparation for electron microscopy, and clarification of lipemic samples.
  • Versatile and Convenient Benchtop Unit
  • Air Turbine-Driven
  • Results within Short Run Times
  • Ideal for Receptor-Binding Studies and Lipoprotein Assays
  • Variety of Rotors
The Airfuge® is air turbine driven and delivers results in a fraction of the time of larger units
The speed is air-pressure regulated and braking is first achieved by coasting, then friction braking by engagement of the brake pin. The rotor operates at a temperature a few degrees above ambient.
The combination of high g-force and short path length results in remarkably short run times to produce results. Runs can be timed from 0–300 minutes in 5 minute increments; a HOLD button can be used for longer runs.
Ordering information: Supplied with a manufacturer's one-year instrument and rotor warranty. Complete with connecting hose and air filter. Rotors sold separately (BK347593 series).